Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant Green, Sigma-Aldrich

Catalog No. 01259078 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-259-078 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-259-078 Supplier MilliporeSigma Supelco Supplier No. B401425G
Add to Cart
Add to Cart

Certified biological stain for microbiological staining, identification, and bacteriostatic use.

Certified for bacteriostatic potency. Tested as a bacteriostatic agent against a spore-former and two members of the colon-typhoid group.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brilliant Green
CAS 633-03-4
Color Dark Green
Quantity 25 g
Molecular Weight (g/mol) 482.63 g/mol
Percent Purity ≥90%
Grade Certified Biological Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.