Learn More
Alkali Scientific Brilliant Green w/Naladixic Acid (100 µg) 20mL Plate, 10/Pk
SDP
Catalog No. 5000662028
Encompass_Preferred
Brilliant Green Agar with Nalidixic Acid is a selective and differential medium used for isolating Salmonella species, excluding Salmonella Typhi, from samples with high background flora. Each sterile 100 × 15 mm plate contains 20 mL of Brilliant Green Agar base supplemented with nalidixic acid to enhance selectivity. The dye brilliant green inhibits gram positive bacteria and many gram negative competitors, while the antibiotic further suppresses organisms such as Proteus and Escherichia coli. Lactose and sucrose fermentation reactions are monitored using phenol red as a pH indicator. Salmonella species generally do not ferment these sugars and form pink to white colonies surrounded by a red medium. This characteristic appearance allows microbiologists to differentiate Salmonella from fermenting organisms during food safety and environmental testing.
Catalog No. 50-006-62028 Supplier Alkali Scientific Supplier No. BGAP00
Brilliant Green Agar with Nalidixic Acid is a selective and differential medium used for isolating Salmonella species, excluding Salmonella Typhi, from samples with high background flora. Each sterile 100 × 15 mm plate contains 20 mL of Brilliant Green Agar base supplemented with nalidixic acid to enhance selectivity. The dye brilliant green inhibits gram positive bacteria and many gram negative competitors, while the antibiotic further suppresses organisms such as Proteus and Escherichia coli. Lactose and sucrose fermentation reactions are monitored using phenol red as a pH indicator. Salmonella species generally do not ferment these sugars and form pink to white colonies surrounded by a red medium. This characteristic appearance allows microbiologists to differentiate Salmonella from fermenting organisms during food safety and environmental testing.