Learn More
MOLTOX Molecular Toxicology Inc Brilliant Green Agar/XLD Aga, Bi-Plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948695
Encompass_Preferred
Brilliant Green Agar is a highly selective medium for the isolation of Salmonella other than S. typhi from materials such as feces. It is used in procedures for the examination of water and wastewater as well as for use in the performance of Microbial Limit Tests. Xylose Lysine Desoxycholate Agar (XLD) is a selective medium for the isolation and differentiation of enteric pathogens, particularly the more fastidious pathogens such as Shigella. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-695 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S86
Brilliant Green Agar is a highly selective medium for the isolation of Salmonella other than S. typhi from materials such as feces. It is used in procedures for the examination of water and wastewater as well as for use in the performance of Microbial Limit Tests. Xylose Lysine Desoxycholate Agar (XLD) is a selective medium for the isolation and differentiation of enteric pathogens, particularly the more fastidious pathogens such as Shigella. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.