Learn More
Agilent Technologies Brilliant II QRT-PCR Master Mix, 1-Step, 400 reactions, 25 μl/reaction, Moloney-based RT, 50°C, 5.5 mM MgCl2, multiplexing: true, storage: -20°C, stability: 6 months
Catalog No. NC2134867
Encompass
Brilliant II QRT-PCR Master Mix, 1-Step, 400 reactions, 25 μl/reaction, Moloney-based RT, 50°C, 5.5 mM MgCl2, multiplexing: true, storage: -20°C, stability: 6 months
This item is not returnable. View return policy
Catalog No. NC2134867 Supplier Agilent Technologies Supplier No. 600809
Brilliant II QRT-PCR Master Mix, 1-Step, 400 reactions, 25 μl/reaction, Moloney-based RT, 50°C, 5.5 mM MgCl2, multiplexing: true, storage: -20°C, stability: 6 months