Learn More
ION BIOSCIENCES Brilliant Thallium Assay pIONeer kit - 2 plates
Supplier:  ION BIOSCIENCES 110002
Encompass_Preferred
ION Biosciences’ Brilliant Thallium (Tl⁺) Assay pIONeer kit is a total assay solution for multi-well plate-based, high-throughput measurements of Tl⁺ flux through K⁺, Na⁺, non-selective cation channels, and some Na⁺ or K⁺ transporters. ION Biosciences Brilliant Thallium Assay excels in potassium channel assays, standing out among market leaders for its robust signal amplitude, high Z’, customizability, and cost-effectiveness. A potassium channel assay, and so much more. ION’s high-throughput, no wash thallium (Tl⁺) flux assay is an optimal solution for measuring the activity of potassium (K⁺), sodium (Na⁺), and non-selective cation channels and their effectors including transporters and GPCRs. The ION Biosciences’ Brilliant Thallium Assay is also useful for a wide range of effectors of ion channels and transporters including G protein-coupled receptors, lipid and protein kinase, for example. 2-plate kit.
Catalog No. NC4043237