Learn More
ION BIOSCIENCES Brilliant Thallium Snapshot Assay Flex kit - 10 plates
Catalog No. NC2866842
Encompass
ION Biosciences' Brilliant Thallium Snapshot Assay Flex kit is an endpoint-compatible potassium channel assay. Brilliant Thallium Snapshot kit is a thallium flux assay adaptation designed to work with standard plate readers, microscopes, flow cytometers, and more, for measuring flux through potassium (K⁺), sodium (Na⁺), and non-selective cation channels, transporters, and their effectors, including GPCRs. 10 plate kit.
Catalog No. NC2866842 Supplier ION BIOSCIENCES Supplier No. 1101110
ION Biosciences' Brilliant Thallium Snapshot Assay Flex kit is an endpoint-compatible potassium channel assay. Brilliant Thallium Snapshot kit is a thallium flux assay adaptation designed to work with standard plate readers, microscopes, flow cytometers, and more, for measuring flux through potassium (K⁺), sodium (Na⁺), and non-selective cation channels, transporters, and their effectors, including GPCRs. 10 plate kit.