Learn More
Biolegend Brilliant Violet 421™ anti-human IgG Fc
Supplier:  Biolegend 410704
Encompass_Preferred
Brilliant Violet 421™ anti-human IgG Fc [M1310G05]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC0991437