Learn More
Biolegend Brilliant Violet 421™ anti-mouse IgG2a
Supplier:  Biolegend 407117
Encompass_Preferred
Brilliant Violet 421™ anti-mouse IgG2a [RMG2a-62]; Isotype: Rat IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1626125