Learn More
Biolegend Brilliant Violet 421™ anti-HA.11 Epitope Tag
Supplier:  Biolegend 682405
Encompass_Preferred
Brilliant Violet 421™ anti-HA.11 Epitope Tag [16B12]; Isotype: Mouse IgG1, κ; Reactivity: YPYDVPDYA Tag; Apps: ICC; Size: 25 ug
Catalog No. NC1610824