Learn More
Biolegend Brilliant Violet 421™ anti-human CD36
Supplier:  Biolegend 336230
Encompass_Preferred
Brilliant Violet 421™ anti-human CD36 [5-271]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1773379