Learn More
Biolegend Brilliant Violet 421™ anti-human Ig light chain κ
Supplier:  Biolegend 316518
Encompass_Preferred
Brilliant Violet 421™ anti-human Ig light chain κ [MHK-49]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. NC1263231