Learn More
Biolegend Brilliant Violet 421™ anti-human CD14
Supplier:  Biolegend 367144
Encompass_Preferred
Brilliant Violet 421™ anti-human CD14 [63D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1925792