Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD4
Supplier:  Biolegend 100543
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD4 [RM4-5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, ICC, IHC-F; Size: 125 uL
Catalog No. NC0233414