Learn More
Biolegend Brilliant Violet 421™ anti-human CD268 (BAFF-R) [11C1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 502527737
Encompass_Preferred
Brilliant Violet 421™ anti-human CD268 (BAFF-R) [11C1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-252-7737 Supplier Biolegend Supplier No. 316931
Brilliant Violet 421™ anti-human CD268 (BAFF-R) [11C1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests