Learn More
Biolegend Brilliant Violet 421TM anti-hu
Supplier:  Biolegend 374211
Encompass_Preferred
Brilliant Violet 421™ anti-human CD86 [BU63]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2069