Learn More
Biolegend Brilliant Violet 421TM anti-hu
Supplier:  Biolegend 348618
Encompass_Preferred
Brilliant Violet 421™ anti-human CD243 (MDR-1) [UIC2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-272-2220