Learn More
Biolegend Brilliant Violet 421TM anti-hu
Catalog No. 502721469
Encompass_Preferred
Brilliant Violet 421™ anti-human IL-9 [MH9A3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-272-1469 Supplier Biolegend Supplier No. 507707
Brilliant Violet 421™ anti-human IL-9 [MH9A3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests