Learn More
Biolegend Brilliant Violet 421TM anti-mo
Catalog No. 502740635
Encompass_Preferred
Brilliant Violet 421™ anti-mouse GM-CSF [MP1-22E9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC; Size: 50 μg
Catalog No. 50-274-0635 Supplier Biolegend Supplier No. 505427
Add to Cart
Add to Cart
Brilliant Violet 421™ anti-mouse GM-CSF [MP1-22E9]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC; Size: 50 μg