Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD25 [A18246A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Supplier:  Biolegend 113705
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD25 [A18246A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. 50-238-8221