Learn More
Biolegend Brilliant Violet 510™ anti-human CD80
Supplier:  Biolegend 305234
Encompass_Preferred
Brilliant Violet 510™ anti-human CD80 [2D10]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1382401