Learn More
Biolegend Brilliant Violet 510™ anti-mouse F4/80
Supplier:  Biolegend 123135
Encompass_Preferred
Brilliant Violet 510™ anti-mouse F4/80 [BM8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC0608989