Learn More
Biolegend Brilliant Violet 510™ anti-mouse CD11c
Supplier:  Biolegend 117338
Encompass_Preferred
Brilliant Violet 510™ anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 500 uL
Catalog No. NC1197252