Learn More
Biolegend Brilliant Violet 510TM anti-hu
Catalog No. 502740681
Encompass_Preferred
Brilliant Violet 510™ anti-human CD61 [VI-PL2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-274-0681 Supplier Biolegend Supplier No. 336434
Brilliant Violet 510™ anti-human CD61 [VI-PL2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests