Learn More
Biolegend Brilliant Violet 510TM anti-hu
Catalog No. 502745672
Encompass_Preferred
Brilliant Violet 510™ anti-human CD66b [6/40c]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-274-5672 Supplier Biolegend Supplier No. 392929
Brilliant Violet 510™ anti-human CD66b [6/40c]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests