Learn More
Biolegend Brilliant Violet 510TM anti-hu
Supplier:  Biolegend 386810
Encompass_Preferred
Brilliant Violet 510™ anti-human CD66b [W20104A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-279-6465