Learn More
Biolegend Brilliant Violet 510TM anti-hu
Supplier:  Biolegend 350025
Encompass_Preferred
Brilliant Violet 510™ anti-human CD134 (OX40) [Ber-ACT35 (ACT35)]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2213