Learn More
Biolegend Brilliant Violet 510TM anti-mo
Supplier:  Biolegend 105839
Encompass_Preferred
Brilliant Violet 510™ anti-mouse CD117 (c-kit) [2B8]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg
Catalog No. 50-272-2785