Learn More
Biolegend Brilliant Violet 510TM anti-mo
Catalog No. 502906292
Encompass_Preferred
Brilliant Violet 510™ anti-mouse CD1d (CD1.1, Ly-38) [1B1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg
Catalog No. 50-290-6292 Supplier Biolegend Supplier No. 123537
Brilliant Violet 510™ anti-mouse CD1d (CD1.1, Ly-38) [1B1]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg