Learn More
Biolegend Brilliant Violet 605™ anti-human CD141 (Thrombomodulin)
Supplier:  Biolegend 344118
Encompass_Preferred
Brilliant Violet 605™ anti-human CD141 (Thrombomodulin) [M80]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 100 tests
Catalog No. NC1622486