Learn More
Biolegend Brilliant Violet 605™ anti-human HLA-A,B,C
Supplier:  Biolegend 311432
Encompass_Preferred
Brilliant Violet 605™ anti-human HLA-A,B,C [W6/32]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cat (Feline), Cattle (Bovine, Cow), Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1744394