Learn More
Biolegend Brilliant Violet 605™ anti-human CD64
Supplier:  Biolegend 305034
Encompass_Preferred
Brilliant Violet 605™ anti-human CD64 [10.1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests
Catalog No. NC1772849