Learn More
Biolegend Brilliant Violet 605™ anti-human CD45RO
Supplier:  Biolegend 304238
Encompass_Preferred
Brilliant Violet 605™ anti-human CD45RO [UCHL1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Chimpanzee, Cynomolgus, Common Marmoset; Apps: FC; Size: 100 tests
Catalog No. NC0564244