Learn More
Biolegend Brilliant Violet 605™ anti-human CD56 (NCAM)
Supplier:  Biolegend 362538
Encompass_Preferred
Brilliant Violet 605™ anti-human CD56 (NCAM) [5.1H11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1386901