Learn More
Biolegend Brilliant Violet 605™ anti-human CD163
Supplier:  Biolegend 333616
Encompass_Preferred
Brilliant Violet 605™ anti-human CD163 [GHI/61]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1490059