Learn More
Biolegend Brilliant Violet 605TM anti-hu
Catalog No. 502722299
Encompass_Preferred
Brilliant Violet 605™ anti-human TCR Vδ2 [B6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2299 Supplier Biolegend Supplier No. 331429
Brilliant Violet 605™ anti-human TCR Vδ2 [B6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests