Learn More
Biolegend Brilliant Violet 605TM anti-hu
Supplier:  Biolegend 306025
Encompass_Preferred
Brilliant Violet 605™ anti-human CD123 [6H6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2446