Learn More
Biolegend Brilliant Violet 605TM anti-hu
Supplier:  Biolegend 304508
Encompass_Preferred
Brilliant Violet 605™ anti-human CD184 (CXCR4) Recombinant [QA18A64]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-286-5932