Learn More
Biolegend Brilliant Violet 605™ anti-mouse CD223 (LAG-3) [C9B7W]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg
Catalog No. 502723449
Encompass_Preferred
Brilliant Violet 605™ anti-mouse CD223 (LAG-3) [C9B7W]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg
Catalog No. 50-272-3449 Supplier Biolegend Supplier No. 125257
Brilliant Violet 605™ anti-mouse CD223 (LAG-3) [C9B7W]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg