Learn More
Biolegend Brilliant Violet 605TM anti-mo
Supplier:  Biolegend 156547
Encompass_Preferred
Brilliant Violet 605™ anti-mouse NK-1.1 [S17016D]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg
Catalog No. 50-274-0729