Learn More
Biolegend Brilliant Violet 650™ anti-mouse CD4
Supplier:  Biolegend 100546
Encompass_Preferred
Brilliant Violet 650™ anti-mouse CD4 [RM4-5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 uL
Catalog No. NC0493814