Learn More
Biolegend Brilliant Violet 650™ anti-mouse/human CD11b
Supplier:  Biolegend 101259
Encompass_Preferred
Brilliant Violet 650™ anti-mouse/human CD11b [M1/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Rhesus, Baboon, Chimpanzee,Rabbit (Lapine); Apps: FC; Size: 50 ug
Catalog No. NC1167698