Learn More
Biolegend Brilliant Violet 650TM anti-
Supplier:  Biolegend 376211
Encompass_Preferred
Brilliant Violet 650™ anti-human TNF-α [W19063E]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-253-7872