Learn More
Biolegend Brilliant Violet 650TM anti-hu
Supplier:  Biolegend 350050
Encompass_Preferred
Brilliant Violet 650™ anti-human CD134 (OX40) [Ber-ACT35 (ACT35)]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-274-0671