Learn More
Biolegend Brilliant Violet 711™ anti-human CD223 (LAG-3)
Supplier:  Biolegend 369320
Encompass_Preferred
Brilliant Violet 711™ anti-human CD223 (LAG-3) [11C3C65]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1411426