Learn More
Biolegend Brilliant Violet 711™ anti-mouse CD103
Supplier:  Biolegend 121435
Encompass_Preferred
Brilliant Violet 711™ anti-mouse CD103 [2E7]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1552667