Learn More
Biolegend Brilliant Violet 711™ anti-human CD13
Supplier:  Biolegend 301722
Encompass_Preferred
Brilliant Violet 711™ anti-human CD13 [WM15]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Chimpanzee, Cotton-topped Tamarin; Apps: FC; Size: 100 tests
Catalog No. NC1302873