Learn More
Biolegend Brilliant Violet 711™ anti-human CD39
Supplier:  Biolegend 328228
Encompass_Preferred
Brilliant Violet 711™ anti-human CD39 [A1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1250131