Learn More
Biolegend Brilliant Violet 711 anti-mouse CD45.2 Antibody, 50 ug
Supplier:  Biolegend 109847
Encompass_Preferred
CD45.2 is an alloantigen of CD45, expressed by Ly5.2 bearing mouse strains (e.g., A, AKR, BALB/c, CBA/Ca, CBA/J, C3H/He, C57BL, C57BR, C57L, C58, DBA/1, DBA/2, NZB, SWR, 129). CD45, a member of the protein tyrosine phosphatase (PTP) family, is a 180-240 kD glycoprotein expressed on all hematopoietic cells except mature erythrocytes and platelets.
Catalog No. NC1673766