Learn More
Biolegend Brilliant Violet 711™ anti-human CD69
Supplier:  Biolegend 310944
Encompass_Preferred
Brilliant Violet 711™ anti-human CD69 [FN50]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1025305