Learn More
Biolegend Brilliant Violet 711™ anti-human CD68 [Y1/82A]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 502723310
Encompass_Preferred
Brilliant Violet 711™ anti-human CD68 [Y1/82A]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-272-3310 Supplier Biolegend Supplier No. 333832
Brilliant Violet 711™ anti-human CD68 [Y1/82A]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests